Rat MPC1 (Mitochondrial Pyruvate Carrier Protein 1) ELISA Kit
Catalog # 77526-294
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Rat
- Target protein:MPC1 (Mitochondrial Pyruvate Carrier Protein 1)
- Size:96 wells
- Sample Type:Tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction
- Sensitivity:0.075 ng/ml
- Regulatory Status:RUO
- Cat. No.:77526-294