Rat Acly (ATP-citRate synthase) ELISA Kit
77526-282
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Rat
- Target protein:Acly (ATP-citRate synthase)
- Size:96 wells
- Sample Type:Tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.63 - 40 ng/ml
- Storage Temperature:2…8 °C
- Application:Metabonomics
- Sensitivity:0.266 ng/ml
- Regulatory Status:RUO
- Cat. No.:77526-282