Pig sC5b-9 (Soluble Terminal Complement Complex C5b-9) ELISA Kit
Catalog # 77525-106
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Pig
- Target protein:sC5b-9 (Soluble Terminal Complement Complex C5b-9)
- Size:96 wells
- Sample Type:Plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.79 - 50 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, Metabolic pathway, Apoptosis, Infection immunity
- Sensitivity:0.32 ng/ml
- Regulatory Status:RUO
- Cat. No.:77525-106