Mouse TNFRSF1A (Tumor Necrosis Factor Receptor Superfamily, Member 1A) ELISA Kit
Catalog # 77519-526
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Mouse
- Target protein:TNFRSF1A (Tumor Necrosis Factor Receptor Superfamily, Member 1A)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:78.13 - 5000 pg/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, CD and Adhesion molecule, Apoptosis
- Sensitivity:28 pg/ml
- Regulatory Status:RUO
- Cat. No.:77519-526