Human MAPt (Microtubule Associated Protein Tau) ELISA Kit
77514-882
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:MAPt (Microtubule Associated Protein Tau)
- Size:96 wells
- Sample Type:Tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Metabolic pathway, Neuro science, Developmental science
- Sensitivity:0.054 ng/ml
- Regulatory Status:RUO
- Cat. No.:77514-882