Mouse PAI1 (Plasminogen Activator Inhibitor 1) ELISA Kit
Catalog # 77514-594
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Mouse
- Target protein:PAI1 (Plasminogen Activator Inhibitor 1)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:31.25 - 2000 pg/ml
- Storage Temperature:2…8 °C
- Application:Metabolic pathway, Hematology
- Sensitivity:12.3 pg/ml
- Regulatory Status:RUO
- Cat. No.:77514-594