Human BDNF (Brain Derived Neurotrophic Factor) ELISA Kit
77512-230
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:BDNF (Brain Derived Neurotrophic Factor)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:31.25 - 2000 pg/ml
- Storage Temperature:2…8 °C
- Application:Cytokine, Infection immunity, Neuro science
- Sensitivity:12.7 pg/ml
- Regulatory Status:RUO
- Cat. No.:77512-230