Dog TRF (Transferrin) ELISA Kit
Catalog # 77512-158
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Dog
- Target protein:TRF (Transferrin)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:3.13 - 200 ng/ml
- Storage Temperature:2…8 °C
- Application:Tumor immunity, Infection immunity, Immune molecule
- Sensitivity:1.12 ng/ml
- Regulatory Status:RUO
- Cat. No.:77512-158