Rat NOX2 (Nicotinamide Adenine Dinucleotide Phosphate Oxidase 2) ELISA Kit
Catalog # 77510-658
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Rat
- Target protein:NOX2 (Nicotinamide Adenine Dinucleotide Phosphate Oxidase 2)
- Size:96 wells
- Sample Type:Tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, Enzyme and Kinase, Metabolic pathway, Tumor immunity
- Sensitivity:0.059 ng/ml
- Regulatory Status:RUO
- Cat. No.:77510-658