Mouse TNFSF12 (Tumor Necrosis Factor Ligand Superfamily, Member 12) ELISA Kit
77528-788
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Mouse
- Target protein:TNFSF12 (Tumor Necrosis Factor Ligand Superfamily, Member 12)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:15.63 - 1000 pg/ml
- Storage Temperature:2…8 °C
- Application:Tumor immunity
- Sensitivity:6.9 pg/ml
- Regulatory Status:RUO
- Cat. No.:77528-788