11-KT ( 11-keto Testosterone) ELISA Kit
77526-952
:
- Assay Type (ELISA with LOV):Competitive
- Target protein:11-KT ( 11-keto Testosterone)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:2 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:156.25 - 10000 pg/ml
- Storage Temperature:2…8 °C
- Application:Endocrinology, Reproductive science, Hormone metabolism
- Sensitivity:15.29 pg/ml
- Regulatory Status:RUO
- Cat. No.:77526-952
- Host: General