Rabbit S100A12 (S100 Calcium Binding Protein A12) ELISA Kit
Catalog # 77525-824
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Rabbit
- Target protein:S100A12 (S100 Calcium Binding Protein A12)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, Infection immunity, Immune molecule
- Sensitivity:0.067 ng/ml
- Regulatory Status:RUO
- Cat. No.:77525-824