PGF2a (Prostaglandin F2 Alpha) ELISA Kit
Catalog # 77524-972
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Competitive
- Target protein:PGF2a (Prostaglandin F2 Alpha)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:2 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:12.5 - 800 pg/ml
- Storage Temperature:2…8 °C
- Application:Metabolic pathway, Cardiovascular biology, Hormone metabolism
- Sensitivity:3.88 pg/ml
- Regulatory Status:RUO
- Cat. No.:77524-972
Specifications
About this item
- Host: General