Human CD163 (Cluster Of Differentiation) ELISA Kit
Catalog # 77519-620
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:CD163 (Cluster Of Differentiation)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.63 - 40 ng/ml
- Storage Temperature:2…8 °C
- Application:CD and Adhesion molecule, Metabolic pathway
- Sensitivity:0.273 ng/ml
- Regulatory Status:RUO
- Cat. No.:77519-620