Human M6PR (Mannose-6-Phosphate Receptor) ELISA Kit
Catalog # 77519-488
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:M6PR (Mannose-6-Phosphate Receptor)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:1.57 - 100 ng/ml
- Storage Temperature:2…8 °C
- Application:CD and Adhesion molecule
- Sensitivity:0.63 ng/ml
- Regulatory Status:RUO
- Cat. No.:77519-488