Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Mouse CASP1 (Caspase 1) ELISA Kit

Mouse CASP1 (Caspase 1) ELISA Kit

96 wells
Catalog # 77518-336
Supplier:   AFG Bioscience
Supplier #:  EK243960
Mouse CASP1 (Caspase 1) ELISA Kit
Catalog # 77518-336
Supplier:  AFG Bioscience
CAS Number:  
Mouse CASP1 (Caspase 1) ELISA Kit
Mouse CASP1 (Caspase 1) ELISA Kit

96 wells

CAS Number:  
Price
$434.94
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Size:
    96 wells
  • Cat. No.:
    77518-336
  • Regulatory Status:
    RUO
  • Sensitivity:
    13.5 pg/ml
  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    CASP1 (Caspase 1)
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
  • Detection Range:
    31.25 - 2000 pg/ml
  • Host:
    Mouse
  • Shelf Life:
    6 months (4 °C); 12 months (–20 °C)
  • Time to Results:
    3.5 h
  • Application:
    Signal transduction, Enzyme and Kinase, Apoptosis, Infection immunity
  • Storage Temperature:
    2…8 °C

Specifications