Human ERAP2 (Endoplasmic Reticulum Aminopeptidase 2) ELISA Kit
77518-300
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:ERAP2 (Endoplasmic Reticulum Aminopeptidase 2)
- Size:96 wells
- Sample Type:Tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.32 - 20 ng/ml
- Storage Temperature:2…8 °C
- Application:Enzyme and Kinase
- Sensitivity:0.128 ng/ml
- Regulatory Status:RUO
- Cat. No.:77518-300