Human NET1 (Neuroepithelial Cell Transforming Gene 1) ELISA Kit
77517-052
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:NET1 (Neuroepithelial Cell Transforming Gene 1)
- Size:96 wells
- Sample Type:Tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.32 - 20 ng/ml
- Storage Temperature:2…8 °C
- Application:Neuro science
- Sensitivity:0.125 ng/ml
- Regulatory Status:RUO
- Cat. No.:77517-052