Rat OC (Osteocalcin) ELISA Kit
Catalog # 77514-208
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Rat
- Target protein:OC (Osteocalcin)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:15.63 - 1000 pg/ml
- Storage Temperature:2…8 °C
- Application:Metabolic pathway, Endocrinology, Hormone metabolism, Bone metabolism
- Sensitivity:6.3 pg/ml
- Regulatory Status:RUO
- Cat. No.:77514-208