Human CR2 (Complement Receptor 2) ELISA Kit
77513-512
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:CR2 (Complement Receptor 2)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:78.13 - 5000 pg/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, CD and Adhesion molecule, Infection immunity, Immune molecule
- Sensitivity:31 pg/ml
- Regulatory Status:RUO
- Cat. No.:77513-512