Human CT1 (Cardiotrophin 1) ELISA Kit
Catalog # 77512-576
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:CT1 (Cardiotrophin 1)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:15.63 - 1000 pg/ml
- Storage Temperature:2…8 °C
- Application:Cytokine, Tumor immunity, Cardiovascular biology
- Sensitivity:5.9 pg/ml
- Regulatory Status:RUO
- Cat. No.:77512-576