Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human PTEN (Phosphatase And Tensin Homolog) ELISA Kit
  77512-180
 :  
undefined
Human PTEN (Phosphatase And Tensin Homolog) ELISA Kit
  77512-180
 :  EK240882
 :  

 

  • Assay Type (ELISA with LOV):
    Sandwich
  • Host:
    Human
  • Target protein:
    PTEN (Phosphatase And Tensin Homolog)
  • Size:
    96 wells
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
  • Time to Results:
    3.5 h
  • Shelf Life:
    6 months (4 °C); 12 months (–20 °C)
  • Detection Range:
    0.32 - 20 ng/ml
  • Storage Temperature:
    2…8 °C
  • Application:
    Tumor immunity
  • Sensitivity:
    0.134 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    77512-180