Human MDC (Macrophage Derived Chemokine) ELISA Kit
Catalog # 77511-784
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:MDC (Macrophage Derived Chemokine)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:78.13 - 5000 pg/ml
- Storage Temperature:2…8 °C
- Application:Cytokine, Infection immunity
- Sensitivity:28 pg/ml
- Regulatory Status:RUO
- Cat. No.:77511-784