Rat MBL1 (Mannose-binding protein A) ELISA Kit
77528-582
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Rat
- Target protein:MBL1 (Mannose-binding protein A)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal Transduction, Cancer
- Sensitivity:0.059 ng/ml
- Regulatory Status:RUO
- Cat. No.:77528-582