Mouse PRKAb1 (Protein Kinase, AMP Activated Beta 1) ELISA Kit
77527-658
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Mouse
- Target protein:PRKAb1 (Protein Kinase, AMP Activated Beta 1)
- Size:96 wells
- Sample Type:Tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Enzyme and Kinase, Metabolic pathway
- Sensitivity:0.055 ng/ml
- Regulatory Status:RUO
- Cat. No.:77527-658