Human sCD40L (Soluble Cluster Of Differentiation 40 Ligand) ELISA Kit
Catalog # 77527-514
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:sCD40L (Soluble Cluster Of Differentiation 40 Ligand)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:15.63 - 1000 pg/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, CD and Adhesion molecule, Apoptosis, Tumor immunity, Infection immunity, Immune molecule
- Sensitivity:6.4 pg/ml
- Regulatory Status:RUO
- Cat. No.:77527-514