Human ACSL4 (Acyl Coenzyme A Synthetase Long Chain Family, Member 4) ELISA Kit
Catalog # 77527-118
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:ACSL4 (Acyl Coenzyme A Synthetase Long Chain Family, Member 4)
- Size:96 wells
- Sample Type:Tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Enzyme and Kinase
- Sensitivity:0.053 ng/ml
- Regulatory Status:RUO
- Cat. No.:77527-118