Rat GLP1 (Glucagon Like Peptide 1) ELISA Kit
77526-158
:
- Assay Type (ELISA with LOV):Competitive
- Host:Rat
- Target protein:GLP1 (Glucagon Like Peptide 1)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:2 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:15.63 - 1000 pg/ml
- Storage Temperature:2…8 °C
- Application:Endocrinology, Cardiovascular biology, Hepatology, Hormone metabolism
- Sensitivity:4.92 pg/ml
- Regulatory Status:RUO
- Cat. No.:77526-158