Rat TFR (Transferrin Receptor) ELISA Kit
77521-466
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Rat
- Target protein:TFR (Transferrin Receptor)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.63 - 40 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, CD and Adhesion molecule, Tumor immunity, Infection immunity
- Sensitivity:0.237 ng/ml
- Regulatory Status:RUO
- Cat. No.:77521-466