Guinea pig IL1a (Interleukin 1 Alpha) ELISA Kit
77521-142
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Guinea pig
- Target protein:IL1a (Interleukin 1 Alpha)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:15.63 - 1000 pg/ml
- Storage Temperature:2…8 °C
- Application:Cytokine, Apoptosis
- Sensitivity:6.6 pg/ml
- Regulatory Status:RUO
- Cat. No.:77521-142