Chicken RBP4 (Retinol Binding Protein 4, Plasma) ELISA Kit
Catalog # 77520-446
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Chicken
- Target protein:RBP4 (Retinol Binding Protein 4, Plasma)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Metabolic pathway, Endocrinology, Hormone metabolism
- Sensitivity:0.071 ng/ml
- Regulatory Status:RUO
- Cat. No.:77520-446