Mouse TLR1 (Toll Like Receptor 1) ELISA Kit
Catalog # 77518-802
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Mouse
- Target protein:TLR1 (Toll Like Receptor 1)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.32 - 20 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, Tumor immunity, Infection immunity, Immune molecule, Pulmonology
- Sensitivity:0.108 ng/ml
- Regulatory Status:RUO
- Cat. No.:77518-802