Mouse MGP (Matrix Gla Protein) ELISA Kit
77518-514
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Mouse
- Target protein:MGP (Matrix Gla Protein)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.47 - 30 ng/ml
- Storage Temperature:2…8 °C
- Application:Metabolic pathway, Hematology, Bone metabolism
- Sensitivity:0.19 ng/ml
- Regulatory Status:RUO
- Cat. No.:77518-514