Human CEACAM1 (Carcinoembryonic Antigen Related Cell Adhesion Molecule 1) ELISA Kit
77515-754
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:CEACAM1 (Carcinoembryonic Antigen Related Cell Adhesion Molecule 1)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:1.57 - 100 ng/ml
- Storage Temperature:2…8 °C
- Application:CD and Adhesion molecule
- Sensitivity:0.57 ng/ml
- Regulatory Status:RUO
- Cat. No.:77515-754