Rat COL4a1 (Collagen Type IV Alpha 1) ELISA Kit
Catalog # 77515-106
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Rat
- Target protein:COL4a1 (Collagen Type IV Alpha 1)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:3.13 - 200 ng/ml
- Storage Temperature:2…8 °C
- Application:Metabolic pathway
- Sensitivity:1.21 ng/ml
- Regulatory Status:RUO
- Cat. No.:77515-106