Rat MUC5AC (Mucin 5 Subtype AC) ELISA Kit
77514-926
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Rat
- Target protein:MUC5AC (Mucin 5 Subtype AC)
- Size:96 wells
- Sample Type:Tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.32 - 20 ng/ml
- Storage Temperature:2…8 °C
- Application:Tumor immunity, Infection immunity, Endocrinology, Pulmonology
- Sensitivity:0.116 ng/ml
- Regulatory Status:RUO
- Cat. No.:77514-926