Human TIMP2 (Tissue Inhibitors Of Metalloproteinase 2) ELISA Kit
77514-050
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:TIMP2 (Tissue Inhibitors Of Metalloproteinase 2)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.94 - 60 ng/ml
- Storage Temperature:2…8 °C
- Application:Tumor immunity
- Sensitivity:0.41 ng/ml
- Regulatory Status:RUO
- Cat. No.:77514-050