Mouse IGF2 (Insulin Like Growth Factor 2) ELISA Kit
77513-942
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Mouse
- Target protein:IGF2 (Insulin Like Growth Factor 2)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:15.63 - 1000 pg/ml
- Storage Temperature:2…8 °C
- Application:Tumor immunity, Endocrinology
- Sensitivity:6.8 pg/ml
- Regulatory Status:RUO
- Cat. No.:77513-942