Mouse MYH1 (Myosin Heavy Chain 1, Skeletal Muscle, Adult) ELISA Kit
77513-642
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Mouse
- Target protein:MYH1 (Myosin Heavy Chain 1, Skeletal Muscle, Adult)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:7.82 - 500 pg/ml
- Storage Temperature:2…8 °C
- Application:Metabolic pathway, Developmental science, Bone metabolism
- Sensitivity:3.2 pg/ml
- Regulatory Status:RUO
- Cat. No.:77513-642