Human GNLY (Granulysin) ELISA Kit
77513-546
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:GNLY (Granulysin)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.24 - 15 ng/ml
- Storage Temperature:2…8 °C
- Application:Tumor immunity, Infection immunity, Immune molecule
- Sensitivity:0.08 ng/ml
- Regulatory Status:RUO
- Cat. No.:77513-546