Human APP (Amyloid Precursor Protein) ELISA Kit
Catalog # 77512-910
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:APP (Amyloid Precursor Protein)
- Size:96 wells
- Sample Type:Serum, plasma, cerebrospinal fluid and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:78.13 - 5000 pg/ml
- Storage Temperature:2…8 °C
- Application:Neuro science
- Sensitivity:30 pg/ml
- Regulatory Status:RUO
- Cat. No.:77512-910