Human HO1 (Heme Oxygenase 1, Decycling) ELISA Kit
Catalog # 77512-394
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:HO1 (Heme Oxygenase 1, Decycling)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.32 - 20 ng/ml
- Storage Temperature:2…8 °C
- Application:Enzyme and Kinase, Metabolic pathway, Cardiovascular biology
- Sensitivity:0.119 ng/ml
- Regulatory Status:RUO
- Cat. No.:77512-394