Rabbit IgG (Immunoglobulin G) ELISA Kit
Catalog # 77511-902
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Competitive
- Host:Rabbit
- Target protein:IgG (Immunoglobulin G)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:2 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:1.57 - 100 μg/ml
- Storage Temperature:2…8 °C
- Application:Infection immunity, Immune molecule, Hematology
- Sensitivity:0.59 μg/ml
- Regulatory Status:RUO
- Cat. No.:77511-902