Human SCUBE3 (Signal peptide, CUB and EGF-like domain-containing protein 3) ELISA Kit
Catalog # 77511-206
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:SCUBE3 (Signal peptide, CUB and EGF-like domain-containing protein 3)
- Size:96 wells
- Sample Type:Tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Cardiovascular
- Sensitivity:0.067 ng/ml
- Regulatory Status:RUO
- Cat. No.:77511-206