Human fFN (Fetal Fibronectin) ELISA Kit
77510-798
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:fFN (Fetal Fibronectin)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:1.57 - 100 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, Infection immunity
- Sensitivity:0.67 ng/ml
- Regulatory Status:RUO
- Cat. No.:77510-798