Order Entry
Puerto Rico
ContactUsLinkComponent
Hamster CTSL (Cathepsin L) ELISA Kit
Hamster CTSL (Cathepsin L) ELISA Kit
Catalog # 77528-376
Supplier:  AFG Bioscience
CAS Number:  
undefined
Hamster CTSL (Cathepsin L) ELISA Kit
Catalog # 77528-376
Supplier:  AFG Bioscience
Supplier Number:  EK248980
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Host:
    Hamster
  • Target protein:
    CTSL (Cathepsin L)
  • Size:
    96 wells
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Time to Results:
    3.5 h
  • Shelf Life:
    6 months (4 °C); 12 months (–20 °C)
  • Detection Range:
    62.5 - 4000 pg/ml
  • Storage Temperature:
    2…8 °C
  • Application:
    Tumor immunity, infection immunity
  • Sensitivity:
    26.2 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    77528-376

Specifications

About this item