Rat ATP (Adenosine Triphosphate) ELISA Kit
Catalog # 77527-728
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Competitive
- Host:Rat
- Target protein:ATP (Adenosine Triphosphate)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:2 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:15.63 - 1000 ng/ml
- Storage Temperature:2…8 °C
- Application:Nutrition metabolism
- Sensitivity:5.27 ng/ml
- Regulatory Status:RUO
- Cat. No.:77527-728