Rat ACVA (Activin A) ELISA Kit
Catalog # 77519-196
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Competitive
- Host:Rat
- Target protein:ACVA (Activin A)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:2 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:15.63 - 1000 pg/ml
- Storage Temperature:2…8 °C
- Application:Endocrinology, reproductive science, hormone metabolism
- Sensitivity:4.66 pg/ml
- Regulatory Status:RUO
- Cat. No.:77519-196