Human a2ML1 (Alpha-2-Macroglobulin Like Protein 1) ELISA Kit
Catalog # 77513-700
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:a2ML1 (Alpha-2-Macroglobulin Like Protein 1)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.79 - 50 ng/ml
- Storage Temperature:2…8 °C
- Application:Infection immunity
- Sensitivity:0.34 ng/ml
- Regulatory Status:RUO
- Cat. No.:77513-700