Rabbit TFPI (Tissue Factor Pathway Inhibitor) ELISA Kit
77512-392
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Rabbit
- Target protein:TFPI (Tissue Factor Pathway Inhibitor)
- Size:96 wells
- Sample Type:plasma
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Metabolic pathway, hematology
- Sensitivity:0.058 ng/ml
- Regulatory Status:RUO
- Cat. No.:77512-392